missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 3 Antibody (GPC3/8148R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20282-100ug
This item is not returnable.
View return policy
Description
Glypican 3 Monoclonal antibody specifically detects Glypican 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Glypican 3 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS | |
| Recombinant fragment (around aa400-550) of human Glypican 3 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
| 2719 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| GPC3/8148R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction