missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glypican 5 Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq, Jul 2008].
Specifications
Specifications
| Antigen | Glypican 5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | GPC5 |
| Gene Accession No. | Q8CAL5 |
| Gene Alias | A230034F01Rik; bA93M14.1; glypican 5; glypican proteoglycan 5; glypican-5; Gpc5; Secreted glypican-5 |
| Gene Symbols | GPC5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?