Learn More
Novus Biologicals™ GM-CSF Antibody (gimsilumab), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28666-1mg
 View more versions of this product
Product Code. 30045073
  • 920.00 € / 1mg
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GM-CSF
Monoclonal
Unconjugated
P04141
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
gimsilumab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
CSF2/ GM-CSF
1 mg
Biologically Active Proteins, Cytokine Research, Immunology
1437
Human
Purified