missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11357-0.1mg
This item is not returnable.
View return policy
Description
GM-CSF Polyclonal antibody specifically detects GM-CSF in Bovine samples. It is validated for Western Blot, ELISA
Specifications
| GM-CSF | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| Recombinant bovine GM-CSF | |
| 0.1 mg | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 1437 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction