missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Antibody (SPM469), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-34774DL594
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| GM-CSF | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CSF2 | |
| Recombinant human GM-CSF protein (Uniprot: P04141) | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| SPM469 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Primate | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction