missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMNN Recombinant Rabbit Monoclonal Antibody (24GB4610)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557029
This item is not returnable.
View return policy
Description
GMNN Recombinant Monoclonal Antibody for Western Blot
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
Specifications
| GMNN | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GMNN | |
| AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291137, 51053 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB4610 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75496 | |
| GMNN | |
| Recombinant protein of human Geminin (aa 110-209). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction