Learn More
Invitrogen™ GMNN Recombinant Rabbit Monoclonal Antibody (24GB4610)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557030
 View more versions of this product
Product Code. 30276492
  • 146.00 € / 20µL
Description
GMNN Recombinant Monoclonal Antibody for Western Blot
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GMNN
Recombinant Monoclonal
Unconjugated
GMNN
AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3
Rabbit
Affinity chromatography
RUO
291137, 51053
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB4610
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75496
GMNN
Recombinant protein of human Geminin (aa 110-209).
20 μL
Primary
Human, Rat