missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GMP Human T Cell Media

Product Code. 30306025
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306025 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30306025 Supplier R&D Systems™ Supplier No. CCM038GMP1B
  • ONLINE EXCLUSIVE PRICE
    369.00 €
    348.60 € / 1 L
    Save 20.40 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For robust T cell expansion with GMP-grade media in xeno-free conditions

/////Phenol Red-Free formulation in 1 L bottles (CCM038-GMP-1L) or 1 L bags (CCM038-GMP-1B)/////Robust T cell expansion under xeno-free conditions/////Optimized for use with Bio-Techne cytokines and G-Rex™ bioreactors/////Compatible with a variety of T cell activation methods/////Supports T cell expansion from PBMCs
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1000 mL
Species Human
Content And Storage Store the unopened product at 2-8 °C and protected from light. Do not use past the use by date on the Certificate of Analysis.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.