Learn More
GNB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10081-1-AP-150UL
 View more versions of this product
Product Code. 16826233
  • 410.00 € / 150µL
Description
Heterotrimeric guanine nucleotide-binding proteins in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.
Specifications
Specifications
GNB3
Polyclonal
Unconjugated
GNB3
GNB3, Transducin beta chain 3
Rabbit
Antigen Affinity Chromatography
RUO
14695, 2784, 60449
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16520, Q61011
GNB3
GNB3 Fusion Protein Ag0127
150 μL
Primary
Human, Mouse, Rat