Learn More
GNRHR Rabbit, anti-Human, Clone: 700000000000, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82847-1-RR-20UL
 View more versions of this product
Product Code. 18009805
  • 149.00 € / 20µL
Description
The GNRHR antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human GNRHR. This antibody recognizes human, mouse antigen. The GNRHR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
GNRHR
700000000000
GnRH R, GnRH receptor, GNRHR, GNRHR1, GRHR, LHRHR, LRHR
20 μL
2798
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa