Learn More
GNRHR Rabbit, anti-Human, Clone: 700000000000, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82847-1-RR-100UL
This item is not returnable.
View return policy
Description
The GNRHR antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human GNRHR. This antibody recognizes human, mouse antigen. The GNRHR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GNRHR | |
| 700000000000 | |
| GnRH R, GnRH receptor, GNRHR, GNRHR1, GRHR, LHRHR, LRHR | |
| 100 μL | |
| 2798 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |