missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GnRHR Recombinant Rabbit Monoclonal Antibody (HL2638)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556726
This item is not returnable.
View return policy
Description
GnRHR Recombinant Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes the receptor for type 1 gonadotropin-releasing hormone. This receptor is a member of the seven-transmembrane, G-protein coupled receptor (GPCR) family. It is expressed on the surface of pituitary gonadotrope cells as well as lymphocytes, breast, ovary, and prostate. Following binding of gonadotropin-releasing hormone, the receptor associates with G-proteins that activate a phosphatidylinositol-calcium second messenger system. Activation of the receptor ultimately causes the release of gonadotropic luteinizing hormone (LH) and follicle stimulating hormone (FSH). Defects in this gene are a cause of hypogonadotropic hypogonadism (HH). Alternative splicing results in multiple transcript variants encoding different isoforms. More than 18 transcription initiation sites in the 5 region and multiple polyA signals in the 3 region have been identified for this gene.
Specifications
| GnRHR | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P30968, P30969, Q01776 | |
| Gnrhr | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human GnRHR. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2638 | |
| Unconjugated | |
| Gnrhr | |
| GH1; GnRH receptor; Gnrhr; gnRH-R; GNRHR1; gonadotropin releasing hormone receptor; gonadotropin releasing hormone receptor type-I; gonadotropin-releasing hormone (type 1) receptor 1; gonadotropin-releasing hormone receptor; GRHR; growth hormone; HH7; HHG; hypergonadotropic hypogonadism; LCGR; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; LGR2; LH/CGR; LH/CG-R; LHCGR; LHR; Lh-rh receptor; Lhrhr; LRHR; LSH-R; luliberin receptor; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; type I GnRH receptor; Type I GNRHR1; ULG5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14715, 2798, 81668 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction