Learn More
Goat anti-Canine IgM Fc Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69669
532.10 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18633358
  • 668.00 € / 1mL
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Canine samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgM Fc
Polyclonal
FITC
Lyophilized from PBS (pH 7.2)
Purified normal IgM isolated from pooled dog serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
B Cell Development and Differentiation Markers, Immunology
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C in the dark.
ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
LYOPH
ELISA, Immunohistochemistry-Frozen, Immunocytochemistry
Goat
Ion exchange chromatography
RUO
Secondary
Canine
Lyophilized