Learn More
ERAP2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ERAP2.
Brand: Abnova PAB11523.100ug
Description
Aminopeptidases hydrolyze N-terminal amino acids of proteins or peptide substrates. Major histocompatibility complex (MHC) class I molecules rely on aminopeptidases such as ERAP1 (MIM 606832) and LRAP to trim precursors to antigenic peptides in the endoplasmic reticulum (ER) following cleavage in the cytoplasm by tripeptidyl peptidase II (TPP2; MIM 190470) (Tanioka et al., 2003 [PubMed 12799365]).[supplied by OMIM
Sequence: C-QYLKKFSYRNAKNDSpecifications
| ERAP2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ERAP2 | |
| FLJ23633/FLJ23701/FLJ23807/L-RAP/LRAP | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 64167 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ERAP2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_071745.1 | |
| ERAP2 | |
| A synthetic peptide corresponding to human ERAP2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only