Learn More
Goat anti-Mouse IgM Fc Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69724
532.10 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18604538
  • 618.00 € / 1mL
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgM Fc
Polyclonal
Biotin
Goat
Ion exchange chromatography
RUO
Secondary
Mouse
Lyophilized
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
LYOPH
Lyophilized from PBS (pH 7.2)
Highly purified homogenous IgM isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
B Cell Development and Differentiation Markers, Immunology
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C in the dark.