missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), R&D Systems™

Product Code. 18210984 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18210984 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18210984 Supplier Novus Biologicals Supplier No. NB7187
  • ONLINE EXCLUSIVE PRICE
    282.37 €
    248.85 € / 1mL
    Save 33.52 €
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Goat Polyclonal Antibody has been used in 3 publications

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000 - 1:30000, ELISA 1:10000 - 1:100000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Host Species Goat
Immunogen Rabbit IgG-heavy and light chain
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with rabbit IgG and with light chains common to other rabbit immunoglobulins. No antibody was detected against nonimmunoglobulin serum proteins. Antiserum was cross adsorbed using a human immunosorbent to remove cross reactive antibodies. Less than 0.1% cross reactivity to human IgG was detected. This antibody may cross react with IgG from other species.
Target Species Rabbit
Content And Storage Store at 4C. Do not freeze.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.