missing translation for 'onlineSavingsMsg'
Learn More

TF, Goat, Polyclonal Antibody, Abnova™

Product Code. 16080947
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16080947 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16080947 Supplier Abnova Supplier No. PAB13847.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Goat polyclonal antibody raised against TF.

This gene encodes a glycoprotein with an approximate molecular weight of 76.5kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq

Specifications

Antigen TF
Applications Immunoassay
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against TF.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM Tris, 500mM NaCl, pH 7.5 (0.05% sodium azide)
Gene TF
Gene Alias DKFZp781D0156/PRO1557/PRO2086
Gene Symbols TF
Host Species Goat
Immunogen Purified human TF.
Quantity 10 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity Monospecific (single arc) by Immunoelectrophoresis (IEP) against normal human plasma and 2x concentrated pooled human serum.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.