missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Complex Antibody (371-4), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34534AF405
This item is not returnable.
View return policy
Description
Golgi Complex Monoclonal specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Golgi Complex | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This monoclonal antibody recognizes an antigen associated with the Golgi complex in human cells only. It can be used to stain the Golgi complex in cell or tissue preparations and can be used as a Golgi marker in subcellular fractions. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells. This monoclonal antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. The Golgi apparatus is an organelle present in all eukaryotic cells that forms a part of the endomembrane system. The primary function of the Golgi apparatus is to process and package macromolecules synthesized by the cell for exocytosis or use within the cell. The Golgi is made up of a stack of flattened, membrane-bound sacs known as cisternae, with three functional regions: the cis face, medial region and trans face. Each region consists of various enzymes that selectively modify the macromolecules passing though them, depending on where they are destined to reside. Several spherical vesicles that have budded off of the Golgi are present surrounding the main cisternae. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 371-4 | |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| SU-DHL-1 large cell lymphoma cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction