missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Complex Antibody (SPM581), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32853-0.02mg
This item is not returnable.
View return policy
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Golgi Complex | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/mL, Flow Cytometry 0.5-1.0 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
| SU-DHL-1 large cell lymphoma cells. | |
| 0.02 mg | |
| Primary | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| SPM581 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction