missing translation for 'onlineSavingsMsg'
Learn More

GOLPH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16858874 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16858874 20 μL 20µL
16848874 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16858874 Supplier Proteintech Supplier No. 191121AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GOLPH3 was initially identified as a peripheral membrane protein localized to the trans-Golgi network, but others reported it to be primarily a mitochondrial protein that regulated the mitochondrial mass through the regulation of the mitochondria-specific phospholipid cardiolipin. GOLPH3 has since been implicated in the target of rapamycin (TOR) signalling pathway. Its overexpression in transfected cells led to and increase in anchorage-independent growth and cell proliferation in vitro. Furthermore, GOLPH3-transfected cells enhanced S6 Kinase activity in response to growth factor stimulation by EGF. Simultaneously, AKT phosphorylation increased in these cells, while these events were abrogated in GOLPH3 siRNA treated cells compared to control cells, indicating the GOLPH3 can enhance signalling through TOR-associated complexes. These results suggest that GOLPH3 is a bona fide oncogene and may be a useful target for therapeutic strategies.

Specifications

Antigen GOLPH3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GOLPH3
Gene Accession No. Q9CRA5, Q9ERE4, Q9H4A6
Gene Alias Coat protein GPP34, Golgi phosphoprotein 3, GOLPH3, GOPP1, GPP34, MIDAS
Gene Symbols GOLPH3
Host Species Rabbit
Immunogen GOLPH3 Fusion Protein Ag5443
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64083, 66629, 78961
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.