missing translation for 'onlineSavingsMsg'
Learn More

Gossypol, MedChemExpress

Product Code. 16476176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476176 Supplier Medchem Express Supplier No. HY13407
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Gossypol binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively.

Gossypol binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively.

Specifications

Chemical Name or Material Gossypol
CAS 303-45-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H30O8
Quantity 100 mg
Synonym BL 193
Solubility Information DMSO : 33.33 mg/mL (64.28 mM; Need ultrasonic)
SMILES O=CC1=C(O)C(O)=C(C(C)C)C2=CC(C)=C(C3=C(C)C=C4C(C(C)C)=C(O)C(O)=C(C=O)C4=C3O)C(O)=C12
Molecular Weight (g/mol) 518.55
Formula Weight 518.55
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.