missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOT2 Recombinant Rabbit Monoclonal Antibody (23GB7985), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553974
This item is not returnable.
View return policy
Description
GOT2 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.
Specifications
| GOT2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GOT2 | |
| AL022787; aspartate aminotransferase 2; aspartate aminotransferase precursor (EC 2.6.1.1); aspartate aminotransferase, mitochondrial; aspartate transaminase 2; ASPATA; FABP-1; FABPpm; FABP-pm; fatty acid-binding protein; FLJ40994; Glutamate oxaloacetate transaminase 2; glutamate oxaloacetate transaminase 2, mitochondrial; Glutamate oxaloacetate transaminase 2, mitochondrial (aspartate aminotransferase 2); glutamatic-oxaloacetic transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2; glutamic-oxaloacetic transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); GOT2; Got-2; KAT4; KATIV; KYAT4; kynurenine aminotransferase 4; Kynurenine aminotransferase IV; kynurenine--oxoglutarate transaminase 4; Kynurenine--oxoglutarate transaminase IV; Maat; mAspAT; mitAAT; mitochondrial aspartate aminotransferase; plasma membrane fatty acid binding protein; plasma membrane-associated fatty acid-binding protein; Transaminase A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14719, 25721, 2806 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB7985 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00505, P00507, P05202 | |
| GOT2 | |
| A synthesized peptide derived from human GOT2 (17-72AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction