missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp100 / Melanosome / PMEL17 / SILV (Melanoma Marker) Mouse, Clone: PMEL/2037, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6490-MSM7-P1
Additional Details : Weight : 0.01000kg
Specifications
gp100 / Melanosome / PMEL17 / SILV (Melanoma Marker) | |
Cytoplasmic | |
PMEL/2037 | |
Unconjugated | |
Mouse | |
90-100kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
6490 | |
IgG1 κ |
Immunohistochemistry, ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human SILV protein fragment (apprx. aa 376-502) | |
SK-MEL-28 cells or Melanoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |