missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 Antibody (GP2/8229R), Novus Biologicals™
Rabbit Monoclonal Antibody
268.00 € - 496.00 €
Specifications
| Antigen | GP2 |
|---|---|
| Clone | GP2/8229R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638844
|
Novus Biologicals
NBP3-23678-100ug |
100 μg |
496.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670105
|
Novus Biologicals
NBP3-23678-20ug |
20 μg |
268.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GP2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
| Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| GP2/8229R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 2813 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title