missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp96/HSP90B1/GRP94 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
218.00 € - 351.00 €
Specifications
| Antigen | gp96/HSP90B1/GRP94 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18180252
|
Novus Biologicals
NBP2-24698SS |
0.025 mg |
218.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769903
|
Novus Biologicals
NBP2-24698 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gp96/HSP90B1/GRP94 Polyclonal specifically detects gp96/HSP90B1/GRP94 in Human, Mouse, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| gp96/HSP90B1/GRP94 | |
| Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Equine, Primate | |
| P14625 | |
| 7184 | |
| A synthetic peptide made to an internal portion of the human GRP94 protein (between amino acids 550-650) [UniProt P14625] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| 94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96) | |
| HSP90B1 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and 72% homologous in mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title