Learn More
GPC2 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27594-1-AP-20UL
This item is not returnable.
View return policy
Description
The GPC2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GPC2. This antibody recognizes mouse, rat antigen. The GPC2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GPC2 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| glypican 2, GPC2 | |
| 20 μL | |
| 221914 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |