missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPHA2 Rabbit anti-Human, Clone: 64, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90250-50UL
This item is not returnable.
View return policy
Description
GPHA2 Monoclonal specifically detects GPHA2 in Human samples. It is validated for ELISA.
Specifications
| GPHA2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 170589 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 64 | |
| ELISA 1:5000-1:10000 | |
| A2, glycoprotein alpha 2, glycoprotein hormone alpha 2, glycoprotein hormone alpha-2, GPA2Thyrostimulin subunit alpha, MGC126572, Putative secreted protein Zsig51, ZSIG51cysteine knot protein | |
| Recombinant Human GPHA2 Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction