missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPM6A Recombinant Rabbit Monoclonal Antibody (HL2473)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556727
This item is not returnable.
View return policy
Description
GPM6A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Neuronal membrane glycoprotein M6-a is a protein that in humans is encoded by the GPM6A gene. Involved in neuronal differentiation, including differentiation and migration of neuronal stem cells. Plays a role in neuronal plasticity and is involved in neurite and filopodia outgrowth, filopodia motility and probably synapse formation. GPM6A-induced filopodia formation involves mitogen-activated protein kinase (MAPK) and Src signaling pathways. May be involved in neuronal NGF-dependent Ca2+ influx. May be involved in regulation of endocytosis and intracellular trafficking of G-protein-coupled receptors (GPCRs); enhances internalization and recycling of mu-type opioid receptor.
Specifications
| GPM6A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35802, Q812E9 | |
| GPM6A | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of mouse GPM6A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2473 | |
| Unconjugated | |
| GPM6A | |
| glycoprotein m6a; GPM6; Gpm6a; M6A; neuronal membrane glycoprotein M6-a; neuronal membrane glycoprotein M6-a-like | |
| Rabbit | |
| Protein A | |
| RUO | |
| 234267, 306439 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction