missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR101 Monoclonal Antibody (909603)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561830
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. Expression of GPR101 has been documented by Northern analysis in human hypothalamus, caudate, and putamen but not in thalamus, hippocampus, frontal cortex, or pons.
Specifications
| GPR101 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q96P66 | |
| GPR101 | |
| NS0 mouse myeloma cell line transfected with human GPR101, Met1-Pro508,. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 909603 | |
| Unconjugated | |
| GPR101 | |
| G protein-coupled receptor 101; G protein-coupled receptor GPR101; Gm365; GPCR6; Gpr101; PAGH2; Probable G-protein coupled receptor 101; RGD1564196 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 83550 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction