missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR110 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15334461
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15334461 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15334461 Supplier Invitrogen™ Supplier No. PA533609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Marmoset (88%).

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR110 (G protein-coupled receptor 110), also known as PGR19, is a 911 amino acid protein that belongs to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Characterized as an adhesion GPCR, GPR110 is a multipass membrane-bound protein with a long amino-terminus that contains multiple domains. One of these domains is the GPCR proteolytic site (GPS), which is essential for proteolytic cleavage of the amino-terminus and for cell surface expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR110
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRF1
Gene Accession No. Q5T601
Gene Alias 5031409J19Rik; Adgrf1; adhesion G protein-coupled receptor F1; adhesion G-protein coupled receptor F1; G protein-coupled receptor 110; G protein-coupled receptor KPG_012; G protein-coupled receptor PGR19; Gpr110; G-protein coupled receptor 110; G-protein coupled receptor KPG_012; hGPCR36; internal gene name of KIRIN laboratory:M5; KPG_009; KPG_012; PGR19; probable G-protein coupled receptor 110; seven transmembrane helix receptor
Gene Symbols ADGRF1
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from N-terminal extracellular domain of human GPR110.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.