missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR126 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81632-1-RR-100UL
This item is not returnable.
View return policy
Description
The GPR126 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human GPR126. This antibody recognizes human, mouse antigen. The GPR126 antibody has been validated for the following applications: IHC, ELISA analysis.Specifications
| GPR126 | |
| Recombinant Monoclonal | |
| 800 μg/mL | |
| APG1; DREG; PS1TP2; VIGR | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Recombinant Antibody | |
| IgG |
| Immunohistochemistry, ELISA | |
| 6M2 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 57211 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering