missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR139 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120407
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, Mouse brain. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: KRFRTMAAAT LKAFFKCQKQ PVQFYTNHNF SITSSPWISP ANSHCIKMLV YQYDKNGKPI KVSP.
Orphan receptor that seems to act through a G(q/11)-mediated pathway.
Specifications
| GPR139 | |
| Polyclonal | |
| Unconjugated | |
| GPR139 | |
| G protein-coupled receptor 139; G protein-coupled receptor PGR3; g(q)-coupled orphan receptor GPRg1; Gm495; Gpr139; GPRG1; G-protein coupled receptor 139; G-protein-coupled receptor PGR3; orphan G protein-coupled receptor TRHRL2; PGR3; Probable G-protein coupled receptor 139 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 124274, 209776 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6DWJ6, Q80UC8 | |
| GPR139 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-353 of human GPR139 (Q6DWJ6). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction