missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR158 Monoclonal Antibody (1027651)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560875
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Orphan receptor.
Specifications
| GPR158 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q5T848 | |
| GPR158 | |
| Human embryonic kidney cell HEK293-derived human GPR158 protein, Ala24-Gln411. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 1027651 | |
| Unconjugated | |
| GPR158 | |
| 5330427M13Rik; G protein-coupled receptor 158; Gpr158; KIAA1136; mKIAA1136; Probable G-protein coupled receptor 158 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 57512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction