Learn More
GPR15L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14716
This item is not returnable.
View return policy
Description
GPR15L Polyclonal specifically detects GPR15L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| C10orf99 | |
| Affinity Purified | |
| IgG |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: KRRPAKAWSGRRTRLCCHRVPSPNSTNLKGHHVRLCKPCKLEPEPRLWVVPGALPQV | |
| 0.1 mL |