missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Antibody (CL10474), Novus Biologicals™
Mouse Monoclonal Antibody
277.00 € - 715.00 €
Specifications
| Antigen | GPR17 |
|---|---|
| Clone | CL10474 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645076
|
Novus Biologicals
NBP3-07993-25ul |
25 μg |
277.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624089
|
Novus Biologicals
NBP3-07993-100ul |
100 μg |
715.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR17 Monoclonal antibody specifically detects GPR17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| GPR17 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: KRLKGPPPSFEGKTNESSLS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL10474 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuroscience | |
| PBS, pH 7.2, containing 40% glycerol | |
| 2840 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title