missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87531
This item is not returnable.
View return policy
Description
GPR17 Polyclonal antibody specifically detects GPR17 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GPR17 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: AEKFRHALCNLLCGKRLKGPPPSFEGKTNESSLSAKSEL | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 2840 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction