missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
370.00 € - 545.00 €
Specifications
| Antigen | GPR27 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18411061
|
Novus Biologicals
NBP1-87003-25ul |
25 μL |
370.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008875
|
Novus Biologicals
NBP1-87003 |
0.1 mL |
545.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR27 Polyclonal specifically detects GPR27 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| GPR27 | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2850 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NRELRDCFRAQFPCCQSPRTTQATHPCDLKGIGL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Unconjugated | |
| RUO | |
| Human | |
| G protein-coupled receptor 27, probable G-protein coupled receptor 27, SREB1Super conserved receptor expressed in brain 1 | |
| GPR27 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title