missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR37 Antibody (S02-9I6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19649-50ul
This item is not returnable.
View return policy
Description
GPR37 Monoclonal antibody specifically detects GPR37 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GPR37 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S02-9I6 | |
| Unconjugated | |
| EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37 | |
| A synthetic peptide of human GPCR GPR37 (Uniprot # O15354) | |
| 50 μg | |
| GPCR | |
| 2861 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction