Learn More
GPR37 Antibody (S02-9I6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19649-100ul
 View more versions of this product
Product Code. 18696483
  • 645.00 € / 100µL
Description
GPR37 Monoclonal antibody specifically detects GPR37 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR37
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Western Blot
S02-9I6
Unconjugated
EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37
A synthetic peptide of human GPCR GPR37 (Uniprot # O15354)
100 μg
GPCR
2861
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.