missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR64 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
292.00 € - 554.00 €
Specifications
| Antigen | GPR64 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Flow Cytometry, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18426560
|
Novus Biologicals
NBP1-84906-25ul |
25ul |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18027785
|
Novus Biologicals
NBP1-84906 |
0.1 mL |
554.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR64 Polyclonal specifically detects GPR64 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GPR64 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, MGC104454, MGC138738, TM7LN2 | |
| ADGRG2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Flow Cytometry, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10149 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SPTVSAPANVNTTSAPPVQTDIVNTSSISDLENQVLQMEKALSLGSLEPNLAGEMINQVSRLLHSPPDMLAPLAQRLLKVVDDIGLQLNFSNTTISLTSPSLALAVIRVNASSFNTTTFVAQDPANLQVSLETQAPE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title