missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR74/NPFFR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-169SS
This item is not returnable.
View return policy
Description
GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| GPR74/NPFFR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NPFFR2 | |
| N terminal sequence MNEKWDTNSSENWHPI and the C terminal sequence ELVMEELKETTNSSEI of the human NPFF2 protein. | |
| 0.025 mg | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 10886 | |
| Human | |
| IgG |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 -1:1000, Immunocytochemistry/Immunofluorescence | |
| GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with the N terminal sequence MNEKWDTNSSENWHPI and the C terminal sequence ELVMEELKETTNSSEI of the human NPFF2 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction