missing translation for 'onlineSavingsMsg'
Learn More

GPRIN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16862154 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16862154 150 μL 150µL
16872154 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16862154 Supplier Proteintech Supplier No. 137711AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects GRIN1 at bands around 110 kDa.

May be involved in neurite outgrowth.

Specifications

Antigen GPRIN1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPRIN1
Gene Accession No. Q3UNH4, Q7Z2K8
Gene Alias GPRIN1, GRIN1, KIAA1893
Gene Symbols GPRIN1
Host Species Rabbit
Immunogen GPRIN1 Fusion Protein Ag4754
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114787, 26913
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.