missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GPT Native Protein

Product Code. 18639037 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1000 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18639037 1000 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18639037 Supplier Novus Biologicals™ Supplier No. NBP262474
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Native Protein

Specific activity - 278 U/mg protein
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 2875
Molecular Weight (g/mol) MolecularWeight-theroretical: 100 kDa
Quantity 1000 U
Storage Requirements Store at -20°C.
Gene Alias AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1
Gene Symbol GPT
Reconstitution Reconstitue with sterile deionised water or dilute buffer.
Research Category metabolism
Purity or Quality Grade >75%
Protein GPT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.