missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPX4 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | GPX4 |
|---|---|
| Concentration | 550 μg/mL |
| Applications | Western Blot, Immunofluorescence, Flow Cytometry (Intracellular Staining), Immunoprecipitation, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237916
|
Proteintech
30388-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236845
|
Proteintech
30388-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GPX4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GPX4. This antibody recognizes human, mouse, rat antigen. The GPX4 antibody has been validated for the following applications: WB, IF-P, FC (Intra), IP, ELISA analysis.Specifications
| GPX4 | |
| Western Blot, Immunofluorescence, Flow Cytometry (Intracellular Staining), Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MCSP; PHGPx; snGPx; snPHGPx | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 550 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 2879 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title