missing translation for 'onlineSavingsMsg'
Learn More

GR148672X, MedChemExpress

Product Code. 16497097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497097 5 mg 5mg
16495276 100 mg 100mg
16480697 50 mg 50mg
16454378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497097 Supplier Medchem Express Supplier No. HY110390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GR148672X is a triacyglycerol hydrolase (TGH) inhibitor with an IC50 of 4 nM extracted from patent WO 2001016358 A2.

GR148672X is a triacyglycerol hydrolase (TGH) inhibitor with an IC50 of 4 nM extracted from patent WO 2001016358 A2.

Specifications

Chemical Name or Material GR148672X
Physical Form Solid
CAS 263890-70-6
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H11F3N2O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 41.67 mg/mL (122.44 mM; Need ultrasonic)
SMILES O=C(C1=CC=CS1)/C(C(C(F)(F)F)=O)=N\NC2=CC=CC(C)=C2
Molecular Weight (g/mol) 340.32
Formula Weight 340.32
Percent Purity 99.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.