Learn More
Granulin Antibody (CL5692) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61424FR
Product Code. 30297013
  • 737.00 € / 0.10mL
Description
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Granulin
Monoclonal
DyLight 680
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin
This antibody was developed using a recombinant protein derived from P28799, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Cycle and Replication, Lipid and Metabolism
2896
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL5692
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified