missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulocyte Marker Antibody (SPM250), Novus Biologicals™
Mouse Monoclonal Antibody
265.00 € - 474.00 €
Specifications
| Antigen | Granulocyte Marker |
|---|---|
| Clone | SPM250 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA 1-5ug/mL for coating, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL, Immunohistochemistry-Frozen 1-2 ug/mL |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18430862
|
Novus Biologicals
NBP2-32856-0.1mg |
0.1 mg |
474.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18430232
|
Novus Biologicals
NBP2-32856-0.02mg |
0.02 mg |
265.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granulocyte Marker Monoclonal antibody specifically detects Granulocyte Marker in Human, Macaque samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Granulocyte Marker | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Macaque | |
| Granulocytes;PML;PMN | |
| IgG1 κ | |
| Protein A or G purified |
| SPM250 | |
| Flow Cytometry 1-2 ug/million cells, ELISA 1-5ug/mL for coating, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL, Immunohistochemistry-Frozen 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes. | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title