missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granulysin Monoclonal Antibody (786343)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561267
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
GNLY is a member of the saposin-like protein (SAPLIP) family and is located in the cytotoxic granules of T cells, which are released upon antigen stimulation. GNLY is present in cytotoxic granules of cytotoxic T lymphocytes and natural killer cells, and it has antimicrobial activity against M. tuberculosis and other organisms.
Specifications
| Granulysin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P22749 | |
| GNLY | |
| Mouse myeloma cell line NS0-derived recombinant human Granulysin, Arg23-Leu145. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA | |
| 786343 | |
| Unconjugated | |
| GNLY | |
| D2S69E; GNLY; Granulysin; LAG2; LAG-2; lymphocyte-activation gene 2; Lymphokine; lymphokine LAG-2; Lymphokine NKG5519; NKG5; Protein NKG5; T-cell activation protein 519; TLA519; T-lymphocyte activation gene 519 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10578 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction