missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody (CL10662), Novus Biologicals™
Mouse Monoclonal Antibody
366.00 € - 625.00 €
Specifications
| Antigen | Granzyme B |
|---|---|
| Clone | CL10662 |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683139
|
Novus Biologicals
NBP3-07984-25ul |
25 μg |
366.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693319
|
Novus Biologicals
NBP3-07984-100ul |
100 μg |
625.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Granzyme B | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TRAVQPLRLPSNKAQVKPGQTCSVAGWGQTAPLGKHSHTLQEVKMTVQEDRKCESDLRHYYDSTIELCVGDPEIKKTSFKGDSGGPLVCNKVAQGIVSYGRNNGMPPRACTKVSS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL10662 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| PBS, pH 7.2, containing 40% glycerol | |
| 3002 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title