missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11894-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist.Specifications
| GRAP2 | |
| Polyclonal | |
| Unconjugated | |
| GRAP2 | |
| Adapter protein GRID, GADS, GRAP 2, GRAP2, GRB 2 like protein, GRB2 related adapter protein 2, GRB2 related adaptor protein 2, GRB2L, GRBLG, GrbX, Grf 40, Grf40, Grf40 adapter protein, GRID, GRPL, Mona, Protein GADS, SH3 SH2 SH3 adapter Mona | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17444, 366962, 9402 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75791, O89100 | |
| Grap2 | |
| GRAP2 Fusion Protein Ag2510 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction